Learn More
S1PR2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 21180-1-AP-20UL
 View more versions of this product
Product Code. 16810805
  • 145.00 € / 20µL
Explore more special offers
Description
Description
The EDG (endothelial differentiation gene) family of G protein coupled receptors consists of eight family members that bind lysophospholipid (LPL) mediators, including sphingosine-1-phosphate (SPP) and lysophosphatidic acid (LPA). EDG-1, EDG-3, EDG-5 (also designated H218 and AGR16) and EDG-8 bind SPP with high-affinity. EDG-6 is a low-affinity receptor for SPP. LPA preferentially binds to EDG-2, EDG-4 and EDG-7. The EDG receptors couple to multiple G proteins to signal through Ras, MAP kinase, Rho, Phospholipase C or other tyrosine kinases, which lead to cell survival, growth, migration and differentiation. EDG-1 signals through Gi proteins to activate Akt and is expressed in glioma cells. EDG-2 is expressed in brain, especially in white matter tract regions, while EDG-3 is expressed in cardiovascular tissue and in cerebellum. EDG-4 is highly expressed on leukocytes and brain, and EDG-5 has wide tissue distribution, including cardiovascular tissue and brain. Expressed in lymphoid and hematopoietic tissues and in lung, EDG-6 signals through Gi/o proteins, which activate growth related pathways.
Specifications
Specifications
S1PR2
Polyclonal
Unconjugated
S1PR2
AGR16, EDG 5, EDG5, Gpcr13, H218, LPB2, S1P receptor 2, S1P receptor Edg 5, S1P2, S1PR2
Rabbit
Antigen Affinity Chromatography
RUO
14739, 29415, 9294
-20°C
Liquid
Immunofluorescence, Western Blot, Immunoprecipitation, Immunocytochemistry
0.21 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O95136, P47752, P52592
S1PR2
S1PR2 Fusion Protein Ag15470
20 μL
Primary
Rat, Mouse, Human