Learn More
Fisherbrand™ S 50 R Ultrasonic Bath
Fisherbrand™ S 50 R Ultrasonic Water Bath features microprocessor-controlled ultrasonic modes and special predefined programs to assist every standard laboratory and cleaning application.
Brand: Fisherbrand™ 1045437
This item is not returnable.
View return policy
Description
- Plastic carrying handles
- Detachable mains cable
- User-friendly clearly arranged operating panel
- Connection rack for laboratory holder, noise-reduction operation
- Ultrasonic tank made of cavitation-resistant stainless steel
- Special efficient and noise-reduced Degas mode (gas liftout effect)
- Turning knob for setting continued and short-period operation from 1 to 30 min
- LED display for both pre-set and actual temperature, and for remaining operating time
Specifications
230 VAC | |
335 x 304 mm (H x Dia.) | |
335 mm | |
Elmasonic S 50 R |
5.3 L | |
230VAC, 150w | |
335 mm | |
150 w |