missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-Phenyl-1,2-ethanediol, 97%, Thermo Scientific Chemicals

25.63 € - 55.78 €

Chemical Identifiers

CAS 25779-13-9
Molecular Formula C8H10O2
Molecular Weight (g/mol) 138.166
MDL Number MFCD00066256
InChI Key PWMWNFMRSKOCEY-MRVPVSSYSA-N
Synonym s-+-1-phenyl-1,2-ethanediol, s-1-phenylethane-1,2-diol, 1s-1-phenylethane-1,2-diol, s-+-1-phenylethane-1,2-diol, s-1-phenyl-1,2-ethanediol, unii-8ib2t1ft6b, 8ib2t1ft6b, 1,2-ethanediol, 1-phenyl-, 1s, styrene glycol, +, r---phenylethylene glycol
PubChem CID 643312
IUPAC Name (1S)-1-phenylethane-1,2-diol
SMILES C1=CC=C(C=C1)C(CO)O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11342287
View Documents
Thermo Scientific Alfa Aesar
L09836.MD
250 mg
25.63 €
250mg
Please to purchase this item. Need a web account? Register with us today!
11332287
View Documents
Thermo Scientific Alfa Aesar
L09836.03
1 g
55.78 €
1g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

25779-13-9
138.166
PWMWNFMRSKOCEY-MRVPVSSYSA-N
643312
C1=CC=C(C=C1)C(CO)O
C8H10O2
MFCD00066256
s-+-1-phenyl-1,2-ethanediol, s-1-phenylethane-1,2-diol, 1s-1-phenylethane-1,2-diol, s-+-1-phenylethane-1,2-diol, s-1-phenyl-1,2-ethanediol, unii-8ib2t1ft6b, 8ib2t1ft6b, 1,2-ethanediol, 1-phenyl-, 1s, styrene glycol, +, r---phenylethylene glycol
(1S)-1-phenylethane-1,2-diol

Specifications

25779-13-9
272°C to 274°C
MFCD00066256
s-+-1-phenyl-1,2-ethanediol, s-1-phenylethane-1,2-diol, 1s-1-phenylethane-1,2-diol, s-+-1-phenylethane-1,2-diol, s-1-phenyl-1,2-ethanediol, unii-8ib2t1ft6b, 8ib2t1ft6b, 1,2-ethanediol, 1-phenyl-, 1s, styrene glycol, +, r---phenylethylene glycol
+39° (c=3 in Ethanol)
(1S)-1-phenylethane-1,2-diol
643312
97%
64°C to 66°C
C8H10O2
3196197
PWMWNFMRSKOCEY-MRVPVSSYSA-N
C1=CC=C(C=C1)C(CO)O
138.166
138.17
(S)-(+)-Phenyl-1,2-ethanediol
Videos
Safety and Handling

Safety and Handling

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.