Learn More
(S)-(-)-Glycidol, 97%, (98% ee), Thermo Scientific Chemicals
35.07 € - 356.78 €
Chemical Identifiers
CAS | 60456-23-7 |
---|---|
Molecular Formula | C3H6O2 |
Molecular Weight (g/mol) | 74.08 |
MDL Number | MFCD00074874 |
InChI Key | CTKINSOISVBQLD-VKHMYHEASA-N |
Synonym | s-glycidol, s-oxiranemethanol, s---glycidol, s-oxiran-2-ylmethanol, s---2,3-epoxy-1-propanol, 2s-oxiran-2-ylmethanol, s-+-glycidol, 2s-oxiran-2-yl methanol, oxiranemethanol, 2sShow More |
PubChem CID | 6973630 |
ChEBI | CHEBI:38690 |
IUPAC Name | [(2S)-oxiran-2-yl]methanol |
SMILES | C1C(O1)CO |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10346884
|
Thermo Scientific Acros
411000010 |
1 g | Glass bottle |
35.07 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10103444
|
Thermo Scientific Acros
411000050 |
5 g | Glass bottle |
103.52 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10224684
|
Thermo Scientific Acros
411000250 |
25 g | Glass bottle |
356.78 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
60456-23-7 | |
74.08 | |
CTKINSOISVBQLD-VKHMYHEASA-N | |
6973630 | |
[(2S)-oxiran-2-yl]methanol |
C3H6O2 | |
MFCD00074874 | |
s-glycidol, s-oxiranemethanol, s---glycidol, s-oxiran-2-ylmethanol, s---2,3-epoxy-1-propanol, 2s-oxiran-2-ylmethanol, s-+-glycidol, 2s-oxiran-2-yl methanol, oxiranemethanol, 2sShow More | |
CHEBI:38690 | |
C1C(O1)CO |
Specifications
60456-23-7 | |
100.0 | |
Colorless to Yellow | |
160°C to 161°C | |
97.5% min. (GC) | |
96% min. (GC) | |
1.4310 to 1.4340 (20°C, 589nm) | |
1.116 | |
s-glycidol, s-oxiranemethanol, s---glycidol, s-oxiran-2-ylmethanol, s---2,3-epoxy-1-propanol, 2s-oxiran-2-ylmethanol, s-+-glycidol, 2s-oxiran-2-yl methanol, oxiranemethanol, 2s | |
CTKINSOISVBQLD-VKHMYHEASA-N | |
[(2S)-oxiran-2-yl]methanol | |
6973630 | |
74.08 | |
Liquid |
96.0 | |
-54°C | |
1.1160g/mL | |
66°C | |
Authentic | |
C3H6O2 | |
MFCD00074874 | |
−12° to −18° (20°C, 589nm) (neat) | |
Solubility in water: soluble | |
C1C(O1)CO | |
74.08 | |
CHEBI:38690 | |
97%,(98% ee) | |
(S)-(-)-Glycidol, 98% ee |
Safety and Handling
GHS H Statement:
Suspected of causing genetic defects.
May cause cancer.
May damage fertility.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.
Toxic if inhaled.
GHS P Statement:
IF exposed or concerned: Get medical advice/attention.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
- (S)-(-)-Glycidol
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Acute toxicity Category 4
- Carcinogenicity Category 1A
- Serious eye damage/eye irritation Category 2
- Germ cell mutagenicity Category 2
- Reproductive toxicity Category 1A
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H350-May cause cancer.
- H360F-May damage fertility.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H341-Suspected of causing genetic defects.
- H331-Toxic if inhaled.
- H335-May cause respiratory irritation.
- H312-Harmful in contact with skin.
- H302-Harmful if swallowed.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.