Learn More
(S)-4-(1-Aminoethyl)phenol, 97%, Thermo Scientific Chemicals
69.85 € - 860.00 €
Chemical Identifiers
CAS | 221670-72-0 |
---|---|
Molecular Formula | C8H11NO |
Molecular Weight (g/mol) | 137.18 |
MDL Number | MFCD03844646 |
InChI Key | CDQPLIAKRDYOCB-UHFFFAOYNA-N |
PubChem CID | 41097924 |
IUPAC Name | 4-[(1S)-1-aminoethyl]phenol |
SMILES | CC(N)C1=CC=C(O)C=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15489878
|
Thermo Scientific Alfa Aesar
H63269.MD |
250 mg | |||||||
15469878
|
Thermo Scientific Alfa Aesar
H63269.03 |
1 g |
230.00 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15479878
|
Thermo Scientific Alfa Aesar
H63269.06 |
5 g |
860.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
(S)-4-(1-Aminoethyl)phenol is used as an intermediate in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications(S)-4-(1-Aminoethyl)phenol is used as an intermediate in organic synthesis.
Notes
Incompatible with oxidizing agents.
Chemical Identifiers
221670-72-0 | |
137.18 | |
CDQPLIAKRDYOCB-UHFFFAOYNA-N | |
4-[(1S)-1-aminoethyl]phenol |
C8H11NO | |
MFCD03844646 | |
41097924 | |
CC(N)C1=CC=C(O)C=C1 |
Specifications
221670-72-0 | |
C8H11NO | |
UN3259 | |
CDQPLIAKRDYOCB-UHFFFAOYNA-N | |
4-[(1S)-1-aminoethyl]phenol | |
41097924 | |
97% | |
(S)-4-(1-Aminoethyl)phenol |
White | |
MFCD03844646 | |
4-1s-1-aminoethyl phenol, s-4-1-aminoethyl phenol, phenol, 4-1s-1-aminoethyl, s-4-1-aminoethyl-phenol, s-4-1-amino-ethyl-phenol, s-4-1-amino-aethyl-phenol, phenol,4-1s-1-aminoethyl, s-4-hydroxy-alpha-methylbenzylamine, phenol, 4-1s-1-aminoethyl-9ci, s-4-hydroxy-? inverted exclamation marka-methylbenzylamine | |
CC(N)C1=CC=C(O)C=C1 | |
137.18 | |
137.18 | |
Solid |
Safety and Handling
GHS H Statement
H301-H314-H318
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302-H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.
missing translation for 'tsca' : No
missing translation for 'storageNote' : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only