missing translation for 'onlineSavingsMsg'
Learn More

(S)-3-Boc-thiazolidine-2-carboxylic acid, 97%, Thermo Scientific Chemicals

Product Code. 15429547
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

(S)-3-Boc-thiazolidine-2-carboxylic acid is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-3-Boc-thiazolidine-2-carboxylic acid is used as an organic chemical synthesis intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 891192-95-3
Molecular Formula C9H15NO4S
Molecular Weight (g/mol) 233.282
MDL Number MFCD02682342
InChI Key HYAXPNDMEODKHI-LURJTMIESA-N
Synonym s-3-boc-2-thiazolidinecarboxylic acid, s-3-tert-butoxycarbonyl thiazolidine-2-carboxylic acid, 2s-3-tert-butoxycarbonyl-1,3-thiazolidine-2-carboxylic acid, 2,3-thiazolidinedicarboxylic acid, 3-1,1-dimethylethyl ester, 2s, s-3-boc-thiazolidine-2-carboxylic acid, n-boc s thiazolidine-2-carboxylic acid, n-boc-s-thiazolidine-2-carboxylic acid, s-thiazolidine-2,3-dicarboxylic acid 3-tert-butyl ester, 2s-3-tert-butoxy carbonyl-1,3-thiazolidine-2-carboxylic acid, 2s-2,3-thiazolidinedicarboxylic acid, 3-1,1-dimethylethyl ester
PubChem CID 14775612
IUPAC Name (2S)-3-[(2-methylpropan-2-yl)oxycarbonyl]-1,3-thiazolidine-2-carboxylic acid
SMILES CC(C)(C)OC(=O)N1CCSC1C(=O)O

Specifications

Melting Point 90°C to 93°C
Quantity 250 mg
Optical Rotation −47° (c=5 in Methanol)
Formula Weight 233.28
Percent Purity 97%
Chemical Name or Material (S)-3-Boc-thiazolidine-2-carboxylic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'