missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-(+)-2-(Anilinomethyl)pyrrolidine, 95%, Thermo Scientific™
86.83 € - 448.00 €
Chemical Identifiers
CAS | 64030-44-0 |
---|---|
Molecular Formula | C11H16N2 |
Molecular Weight (g/mol) | 176.26 |
InChI Key | MCHWKJRTMPIHRA-NSHDSACASA-N |
Synonym | s-+-2-anilinomethyl pyrrolidine, s-n-pyrrolidin-2-ylmethyl aniline, s-2-anilinomethyl pyrrolidine, s-+-n-2-pyrrolidinomethyl aniline, s-phenyl-pyrrolidin-2-ylmethyl-amine, n-2s-pyrrolidin-2-ylmethyl aniline, pubchem8116, 2s-2-anilinomethyl pyrrolidine, s-2-phenylaminomethyl-pyrrolidine, n-2s-pyrrolidin-2-yl methyl aniline |
PubChem CID | 6950385 |
IUPAC Name | N-[[(2S)-pyrrolidin-2-yl]methyl]aniline |
SMILES | C1CC(NC1)CNC2=CC=CC=C2 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11956761
|
Thermo Scientific Acros
445800010 |
1 g |
86.83 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11966761
|
Thermo Scientific Acros
445800050 |
5 g | N/A | ||||||
Description
Chemical Identifiers
64030-44-0 | |
176.26 | |
s-+-2-anilinomethyl pyrrolidine, s-n-pyrrolidin-2-ylmethyl aniline, s-2-anilinomethyl pyrrolidine, s-+-n-2-pyrrolidinomethyl aniline, s-phenyl-pyrrolidin-2-ylmethyl-amine, n-2s-pyrrolidin-2-ylmethyl aniline, pubchem8116, 2s-2-anilinomethyl pyrrolidine, s-2-phenylaminomethyl-pyrrolidine, n-2s-pyrrolidin-2-yl methyl aniline | |
N-[[(2S)-pyrrolidin-2-yl]methyl]aniline |
C11H16N2 | |
MCHWKJRTMPIHRA-NSHDSACASA-N | |
6950385 | |
C1CC(NC1)CNC2=CC=CC=C2 |
Specifications
64030-44-0 | |
126°C (0.1 kPa) | |
C11H16N2 | |
s-+-2-anilinomethyl pyrrolidine, s-n-pyrrolidin-2-ylmethyl aniline, s-2-anilinomethyl pyrrolidine, s-+-n-2-pyrrolidinomethyl aniline, s-phenyl-pyrrolidin-2-ylmethyl-amine, n-2s-pyrrolidin-2-ylmethyl aniline, pubchem8116, 2s-2-anilinomethyl pyrrolidine, s-2-phenylaminomethyl-pyrrolidine, n-2s-pyrrolidin-2-yl methyl aniline | |
C1CC(NC1)CNC2=CC=CC=C2 | |
176.26 | |
176.26 | |
(S)-(+)-2-(Anilinomethyl)pyrrolidine |
19°C | |
95% | |
+ 1.63 | |
MCHWKJRTMPIHRA-NSHDSACASA-N | |
N-[[(2S)-pyrrolidin-2-yl]methyl]aniline | |
6950385 | |
95% |
Safety and Handling
GHS Signal Word: Danger
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- (S)-(+)-2-(Anilinomethyl)pyrrolidine
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title