missing translation for 'onlineSavingsMsg'
Learn More

RUVBL2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

145.00 € - 410.00 €

Specifications

Antigen RUVBL2
Concentration 0.21 mg/mL
Applications Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot, Immunoprecipitation, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16886413
View Documents Promotion Details
Proteintech
10195-1-AP-20UL
20 μL
145.00 €
20µL
Please to purchase this item. Need a web account? Register with us today!
16876413
View Documents Promotion Details
Proteintech
10195-1-AP-150UL
150 μL
410.00 €
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Possesses single-stranded DNA-stimulated ATPase and ATP-dependent DNA helicase (5′ to 3′) activity. Component of the NuA4 histone acetyltransferase complex which is involved in transcriptional activation of select genes principally by acetylation of nucleosomal histones H4 and H2A. This modification may both alter nucleosome - DNA interactions and promote interaction of the modified histones with other proteins which positively regulate transcription. This complex may be required for the activation of transcriptional programs associated with oncogene and proto-oncogene mediated growth induction, tumor suppressor mediated growth arrest and replicative senescence, apoptosis, and DNA repair. The NuA4 complex ATPase and helicase activities seem to be, at least in part, contributed by the association of RUVBL1 and RUVBL2 with EP400. NuA4 may also play a direct role in DNA repair when recruited to sites of DNA damage. RUVBL2 plays an essential role in oncogenic transformation by MYC and also modulates transcriptional activation by the LEF1/TCF1-CTNNB1 complex. May also inhibit the transcriptional activity of ATF2.
Specifications

Specifications

RUVBL2
Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot, Immunoprecipitation, Immunofluorescence
Unconjugated
Rabbit
Mouse, Rat, Human
Q9WTM5, Q9Y230
10856, 20174, 292907
RUVBL2 Fusion Protein Ag0253
Primary
-20°C
RUVBL2
0.21 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
48 kDa TBP interacting protein, CGI 46, ECP 51, ECP51, INO80 complex subunit J, INO80J, Repressing pontin 52, REPTIN, Reptin 52, RuvB like 2, RuvB like 2 (E. coli), RUVBL2, RVB2, TAP54 beta, TIH2, TIP48, TIP49B
Ruvbl2
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications