missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rutin Hydrate, 97+%, Thermo Scientific Chemicals
37.70 € - 512.00 €
Chemical Identifiers
CAS | 207671-50-9 |
---|---|
Molecular Formula | C27H30O16 |
Molecular Weight (g/mol) | 610.52 |
MDL Number | MFCD01319140 |
InChI Key | IKGXIBQEEMLURG-NVPNHPEKSA-N |
Synonym | rutin, rutoside, phytomelin, quercetin 3-rutinoside, birutan, eldrin, myrticolorin, venoruton, 3-rutinosyl quercetin, bioflavonoid |
PubChem CID | 5280805 |
ChEBI | CHEBI:28527 |
IUPAC Name | 2-(3,4-dihydroxyphenyl)-5,7-dihydroxy-3-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-[[(2R,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxymethyl]oxan-2-yl]oxychromen-4-one |
SMILES | C[C@@H]1O[C@@H](OC[C@H]2O[C@@H](OC3=C(OC4=CC(O)=CC(O)=C4C3=O)C3=CC(O)=C(O)C=C3)[C@H](O)[C@@H](O)[C@@H]2O)[C@H](O)[C@H](O)[C@H]1O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10071480
|
Thermo Scientific Acros
132390050 |
5 g | Glass Bottle |
37.70 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10356970
|
Thermo Scientific Acros
132391000 |
100 g | Plastic bottle |
168.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10327020
|
Thermo Scientific Acros
132395000 |
500 g | Plastic bottle |
512.00 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
207671-50-9 | |
610.52 | |
IKGXIBQEEMLURG-NVPNHPEKSA-N | |
5280805 | |
2-(3,4-dihydroxyphenyl)-5,7-dihydroxy-3-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-[[(2R,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxymethyl]oxan-2-yl]oxychromen-4-one |
C27H30O16 | |
MFCD01319140 | |
rutin, rutoside, phytomelin, quercetin 3-rutinoside, birutan, eldrin, myrticolorin, venoruton, 3-rutinosyl quercetin, bioflavonoid | |
CHEBI:28527 | |
C[C@@H]1O[C@@H](OC[C@H]2O[C@@H](OC3=C(OC4=CC(O)=CC(O)=C4C3=O)C3=CC(O)=C(O)C=C3)[C@H](O)[C@@H](O)[C@@H]2O)[C@H](O)[C@H](O)[C@H]1O |
Specifications
207671-50-9 | |
White | |
Authentic | |
C27H30O16 | |
15, 8438 | |
Solubility in water: 1g/8l. Other solubilities: soluble in alkaline solutions,soluble in pyridine and formamide,slightly soluble in alcohol and acetone,practically insoluble in chloroform,carbone,disulfide and benzene | |
C[C@@H]1O[C@@H](OC[C@H]2O[C@@H](OC3=C(OC4=CC(O)=CC(O)=C4C3=O)C3=CC(O)=C(O)C=C3)[C@H](O)[C@@H](O)[C@@H]2O)[C@H](O)[C@H](O)[C@H]1O | |
610.52 | |
CHEBI:28527 | |
97+% | |
Rutin hydrate, 97.0-102.0% |
195.0°C | |
5.5 % to 9.0 % (500mg, 130°C, 3 hrs) (vacuum) | |
97+% | |
MFCD01319140 | |
rutin, rutoside, phytomelin, quercetin 3-rutinoside, birutan, eldrin, myrticolorin, venoruton, 3-rutinosyl quercetin, bioflavonoid | |
IKGXIBQEEMLURG-NVPNHPEKSA-N | |
2-(3,4-dihydroxyphenyl)-5,7-dihydroxy-3-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-[[(2R,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxymethyl]oxan-2-yl]oxychromen-4-one | |
5280805 | |
664.58 | |
Powder |
Safety and Handling
LIGHT SENSITIVE
HYGROSCOPIC
missing translation for 'einecsNumber' : 205-814-1
missing translation for 'rtecsNumber' : VM2975000
missing translation for 'tsca' : TSCA
RUO – Research Use Only