missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ruthenium, 5% on activated carbon powder, reduced, nominally 50% water wet, Thermo Scientific Chemicals
61.50 € - 598.00 €
Chemical Identifiers
CAS | 7440-18-8 |
---|---|
Molecular Formula | Ru |
Molecular Weight (g/mol) | 101.07 |
MDL Number | MFCD00011207 MFCD03458417 |
InChI Key | KJTLSVCANCCWHF-UHFFFAOYSA-N |
Synonym | on carbon, rutenio, black, unii-7ui0tkc3u5, 7ui0tkc3u5, ruthenium, powder, alumina, atom, rutherium black, ruthenium/carbon |
PubChem CID | 23950 |
ChEBI | CHEBI:30682 |
IUPAC Name | ruthenium |
SMILES | [Ru] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11408470
|
Thermo Scientific Alfa Aesar
044338.06 |
5 g |
61.50 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11428470
|
Thermo Scientific Alfa Aesar
044338.14 |
25 g |
199.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11418470
|
Thermo Scientific Alfa Aesar
044338.22 |
100 g |
598.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Ruthenium, 5% on activated carbon powde is used in solar cells, which turn light energy into electrical energy.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7440-18-8 | |
101.07 | |
KJTLSVCANCCWHF-UHFFFAOYSA-N | |
23950 | |
ruthenium |
Ru | |
MFCD00011207 MFCD03458417 | |
on carbon, rutenio, black, unii-7ui0tkc3u5, 7ui0tkc3u5, ruthenium, powder, alumina, atom, rutherium black, ruthenium/carbon | |
CHEBI:30682 | |
[Ru] |
Specifications
7440-18-8 | |
Ru | |
on carbon, rutenio, black, unii-7ui0tkc3u5, 7ui0tkc3u5, ruthenium, powder, alumina, atom, rutherium black, ruthenium/carbon | |
KJTLSVCANCCWHF-UHFFFAOYSA-N | |
ruthenium | |
23950 | |
Odorless |
Powder | |
MFCD00011207 MFCD03458417 | |
Insoluble in water. | |
[Ru] | |
101.07 | |
CHEBI:30682 | |
Ruthenium, 5% on activated carbon powder, Reduced, nominally 50% water wet |
Safety and Handling
missing translation for 'einecsNumber' : 231-127-1
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only