missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Uvex™ Rubiflex S NB80S Chemical Protection Gloves
Cotton interlock supporting material, along with NBR exterior, makes these gloves suitable for applications that require protection against chemicals and mechanical hazards.
49.80 € - 52.05 €
Specifications
Coating Material | NBR |
---|---|
Color | Green |
EN388 Standard | 2121X |
EN ISO 374-1 | Type B |
EN ISO 374-1 CHEMICAL LETTER CODES | JKOPT |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
15376952
|
Uvex™
6019001 |
9 |
49.80 €
Each |
Estimated Shipment: 14-01-2025
Log in to see stock. |
|||||
15356952
|
Uvex™
6019002 |
10 |
49.80 €
Each |
Estimated Shipment: 14-01-2025
Log in to see stock. |
|||||
15366952
|
Uvex™
6019003 |
11 |
52.05 €
Each |
Estimated Shipment: 14-01-2025
Log in to see stock. |
|||||
Description
- Long NBR chemical protection gloves reinforced cotton interlock carrier material
- NBR coating provides mechanical abrasion resistance
- High-quality interlock cotton supporting material increases wearer comfort
- Gloves are resistant to a wide variety of chemicals, acids, alkalis, mineral oils, and solvents
- Ergonomic fit with improved dexterity and instinct
- Ideal for applications in the chemical industry, sewer construction work, and city cleaning
Specifications
NBR | |
2121X | |
JKOPT | |
Interlock Cotton | |
80 cm |
Green | |
Type B | |
Gauntlet | |
Rubiflex S NB80S |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title