missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Uvex™ Rubiflex S NB35S Chemical Protection Gloves
Cotton interlock supporting material, along with NBR exterior, makes these gloves suitable for applications that require protection against chemicals and mechanical hazards.
16.70 €
Specifications
Coating Material | NBR |
---|---|
Color | Green |
Hand | Specific |
EN ISO 374-1 | Type A |
EN ISO 374-1 CHEMICAL LETTER CODES | JKNOPT |
Description
- Long NBR chemical protection gloves reinforced cotton interlock carrier material
- NBR coating provides mechanical abrasion resistance
- High-quality interlock cotton supporting material increases wearer comfort
- Gloves are resistant to a wide variety of chemicals, acids, alkalis, mineral oils, and solvents
- Ergonomic fit with improved dexterity and instinct
- Ideal for applications in the chemical industry, sewer construction work, and city cleaning
Specifications
NBR | |
Specific | |
JKNOPT | |
Interlock Cotton | |
35 cm |
Green | |
Type A | |
Gauntlet | |
Rubiflex S NB35S |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title