Learn More
Rubidium sulfate, 99.8% (metals basis), Thermo Scientific Chemicals
48.00 € - 167.00 €
Chemical Identifiers
CAS | 7488-54-2 |
---|---|
Molecular Formula | H2O4RbS |
Molecular Weight (g/mol) | 183.54 |
MDL Number | MFCD00011190 |
InChI Key | XNNUAZGCOYTNQE-UHFFFAOYSA-N |
PubChem CID | 197088 |
SMILES | [Rb].OS(O)(=O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11380719
|
Thermo Scientific Alfa Aesar
010567.09 |
10 g |
48.00 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11390719
|
Thermo Scientific Alfa Aesar
010567.18 |
50 g |
167.00 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Rubidium sulfate is used in hematology stain.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7488-54-2 | |
183.54 | |
XNNUAZGCOYTNQE-UHFFFAOYSA-N | |
[Rb].OS(O)(=O)=O |
H2O4RbS | |
MFCD00011190 | |
197088 |
Specifications
1050°C | |
Powder | |
MFCD00011190 | |
Soluble in water. | |
[Rb].OS(O)(=O)=O | |
183.54 | |
267 | |
Odorless | |
3.613 g/mL | |
Rubidium sulfate |
7488-54-2 | |
H2O4RbS | |
rubidium sulfate, rubidium sulphate, dirubidium sulfate, rubidium sulfate rb2so4, sulfuric acid, dirubidium salt, rubidium sulfate rb2 so4, dirubidium 1+ sulfate, rubidiumsulfat, sulfuric acid, rubidium salt 1:2, rubidium 1+ sulfate | |
XNNUAZGCOYTNQE-UHFFFAOYSA-N | |
rubidium(1+);sulfate | |
197088 | |
99.8% | |
1.513 | |
(metals basis) |
Seguridad y manipulación
GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 231-301-7
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only