missing translation for 'onlineSavingsMsg'
Learn More

Rubidium nitrate, Puratronic™, 99.975% (metals basis), Thermo Scientific Chemicals

Product Code. 11307007
Click to view available options
Quantity:
1 g
10 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Rubidium nitrate is used in infrared radiation producing pyrotechnic compositions. It acts as a colorant and an oxidizer especially in decoys and illumination flares. Further, it is employed as a starting material in the preparation of other rubidium compounds and rubidium metal, which is utilized for the preparation of catalysts and in scintillation counters. In addition to this, it is used in fireworks to produce a red-violet color.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13126-12-0
Molecular Formula NO3Rb
Molecular Weight (g/mol) 147.47
MDL Number MFCD00011193
InChI Key RTHYXYOJKHGZJT-UHFFFAOYSA-N
Synonym rubidium nitrate, nitric acid, rubidium salt, nitric acid, rubidium salt 1:1, rubidium 1+ nitrate, rubidium 1+ nitronate, acmc-1c2ua, rubidium 1+ ion nitronate, rubidium nitrate, puratronic, rubidium nitrate-rb, rubidium 1+ ion nitrate
PubChem CID 25731
SMILES [Rb+].[O-][N+]([O-])=O

Specifications

Melting Point 310°C
Physical Form Crystalline
Quantity 50 g
UN Number UN1477
Solubility Information Soluble in water and nitric acid. Slightly soluble in acetone.
IUPAC Name rubidium(1+);nitrate
Formula Weight 147.47
Percent Purity 99.975%
Grade Puratronic™
Refractive Index 1.52
Sensitivity Hygroscopic
Density 3.11 g/mL
Assay (metals basis)
Chemical Name or Material Rubidium nitrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'