missing translation for 'onlineSavingsMsg'
Learn More

Rubidium fluoride, Puratronic™, 99.975% (metals basis), Thermo Scientific Chemicals

103.52 € - 371.85 €

Chemical Identifiers

CAS 13446-74-7
Molecular Formula FRb
Molecular Weight (g/mol) 104.466
MDL Number MFCD00011191
InChI Key AHLATJUETSFVIM-UHFFFAOYSA-M
Synonym rubidium fluoride, rubidium fluoride rbf, rubidium monofluoride, rubidium 1+ fluoride, rubidium 1+ ion fluoride, rubidium fluoride, anhydrous, unii-63564tnj4r, acmc-20ajsd, rubidium i fluoride, rubidium fluoride, rbf
PubChem CID 83473
IUPAC Name rubidium(1+);fluoride
SMILES [F-].[Rb+]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11337607
SDS View Documents
Thermo Scientific Alfa Aesar
012873.04
2 g
103.52 €
2g
Please to purchase this item. Need a web account? Register with us today!
11327607
SDS View Documents
Thermo Scientific Alfa Aesar
012873.09
10 g
371.85 €
10g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

As raw materials for preparation of rubidium metal and various rubidium salts, for the manufacturing of catalyst and for the manufacturing of high energy density micro cells and crystal scintillation counters.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H331-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P301+P310-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.

missing translation for 'rtecsNumber' : VL8740000

missing translation for 'tsca' : TSCA

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Rubidium fluoride, Puratronic™, 99.975% (metals basis), Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.