missing translation for 'onlineSavingsMsg'
Learn More

Rubidium fluoride hydrate, 99%, Thermo Scientific™

36.00 € - 157.00 €

Chemical Identifiers

CAS 16422-67-6
Molecular Formula FH2ORb
Molecular Weight (g/mol) 122.481
MDL Number MFCD00149834
InChI Key CTSZMVONLYPOKW-UHFFFAOYSA-M
Synonym rubidium fluoride hydrate, f.rb.h2o, acmc-1c7r7, rubidium fluoride,hydrate 8ci, rubidium fluoride-water 1/1/1, rubidium 1+ ion hydrate fluoride
PubChem CID 57370370
IUPAC Name rubidium(1+);fluoride;hydrate
SMILES O.[F-].[Rb+]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11482868
SDS View Documents
Thermo Scientific Alfa Aesar
A16846.09
10 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11492868
SDS View Documents
Thermo Scientific Alfa Aesar
A16846.18
50 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Raw material used in the perparation of rubidium metal.

Solubility
Soluble in water.

Notes
Store in a cool, dry, conditions in well sealed container.
Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H331-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P260-P264b-P270-P271-P280-P284-P301+P310-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P361-P363-P501c

H301+H311-H315-H319-H330-H335

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.

missing translation for 'einecsNumber' : 236-603-2

missing translation for 'rtecsNumber' : VL8740000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Rubidium fluoride hydrate, 99%, Thermo Scientific™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.