Learn More
Abeba™ Rubber 5250 Shoes
ESD dissipative shoe is acid and base resistant.
Brand:  Abeba™ 5250/41
83.19 EUR valid until 2025-02-13
Use promo code "24130" to get your promotional price.
 View more versions of this product
Product Code. 15336711
  • 85.65 € / 1 pair
Estimated Shipment: 29-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • CE EN ISO 20347:2012 OB A, FO SRC
  • Smooth leather
  • Dark blue
  • PU midsole
  • Slip resistance special rubber outsole sole
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Heel strap foldable, adjustable
  • Leather top layer
Specifications
Specifications
CE EN ISO 20347:2012 OB A, FO SRC
Leather
Slip-resistant
5250
Blue
41
Rubber