missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RRS1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00 € - 410.00 €
Specifications
Antigen | RRS1 |
---|---|
Concentration | 0.32 mg/mL |
Applications | Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16834464
|
Proteintech
15329-1-AP-20UL |
20 μL |
145.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16824464
|
Proteintech
15329-1-AP-150UL |
150 μL |
410.00 €
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Involved in ribosome biogenesis. .Specifications
RRS1 | |
Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Mouse, Human, Rat | |
A1A5P2, Q15050, Q9CYH6 | |
23212, 297784, 59014 | |
RRS1 Fusion Protein Ag7030 | |
Primary | |
-20°C | |
RRS1 |
0.32 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
KIAA0112, RRR, RRS1 | |
Rrs1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title