missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Round magnetic stirring bars with tapered ends have naturally centered pivot point, eliminating need for separate pivot ring
- Smooth surface and slightly raised ends on these bars facilitate efficient movement through solutions
- End diameters are 1mm less than center diameter
- Alnico magnet encapsulated in FDA Grade, USP Class VI Teflon™ PTFE low friction coating; durable and inert
- Manufactured in registered ISO 9001:2008 facility, as verified by SGS certification
- Autoclavable
Certifications
ISO 9001:2008, FDA
Specifications
FDA, ISO 9001:2008 | |
0.35in. | |
9 mm | |
Alnico Magnet |
White | |
1.97in. | |
50 mm | |
Round with tapered ends |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction