missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Rotor Adapters for Rotor S-4x1000

For use with a variety of tubes and plates in different shapes and sizes.

Brand:  Eppendorf™ 5920715003

 View more versions of this product

Product Code. 15728389

  • 329.00 € / Pack of 2

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Adapters for Rotors. Available for use with:
  • 5.5 to 12mL, set of 2 pcs.
  • 50mL, set of 2 pcs.
  • Eppendorf Tubes 5.0mL and 15mL, set of 2 pcs.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

25 mL, 50 mL
For Rotor S-4x1000 high-capacity buckets
13
9 mL
30mm
Conical bottom
13/52
Set of 2 pieces
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers