Learn More
Thermo Scientific Chemicals Rose Bengal, 85%, pure, certified
75.20 € - 149.00 €
Chemical Identifiers
CAS | 632-69-9 |
---|---|
Molecular Formula | C20H4Cl4I4Na2O5 |
Molecular Weight (g/mol) | 1019.644 |
MDL Number | MFCD00005043 |
InChI Key | DPBQSRZQYAYDGY-UHFFFAOYSA-N |
Synonym | Acid Red 94, C.I. 45440 |
PubChem CID | 87244310 |
IUPAC Name | sodium;4,5,6,7-tetrachloro-3',6'-dihydroxy-2',4',5',7'-tetraiodospiro[2-benzofuran-3,9'-xanthene]-1-one |
SMILES | C1=C2C(=C(C(=C1I)O)I)OC3=C(C(=C(C=C3C24C5=C(C(=C(C(=C5Cl)Cl)Cl)Cl)C(=O)O4)I)O)I.[Na].[Na] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10391541
|
Thermo Scientific Chemicals
419150250 |
25 g | Glass bottle |
75.20 €
25g |
Estimated Shipment: 18-11-2024
Log in to see stock. |
||||
10761244
|
Thermo Scientific Chemicals
419151000 |
100 g | Glass bottle |
149.00 €
100g |
In Stock
Log in to see stock. |
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
IndicatorChemical Identifiers
632-69-9 | |
1019.644 | |
DPBQSRZQYAYDGY-UHFFFAOYSA-N | |
87244310 | |
C1=C2C(=C(C(=C1I)O)I)OC3=C(C(=C(C=C3C24C5=C(C(=C(C(=C5Cl)Cl)Cl)Cl)C(=O)O4)I)O)I.[Na].[Na] |
C20H4Cl4I4Na2O5 | |
MFCD00005043 | |
Acid Red 94, C.I. 45440 | |
sodium;4,5,6,7-tetrachloro-3',6'-dihydroxy-2',4',5',7'-tetraiodospiro[2-benzofuran-3,9'-xanthene]-1-one |
Specifications
632-69-9 | |
80% min. | |
C20H4Cl4I4Na2O5 | |
15, 8393 | |
Solubility in water: soluble (36%). | |
Authentic | |
sodium;4,5,6,7-tetrachloro-3',6'-dihydroxy-2',4',5',7'-tetraiodospiro[2-benzofuran-3,9'-xanthene]-1-one | |
87244310 | |
1017.65 | |
Pure | |
>92°C | |
Rose Bengal |
1.21 to 1.95 | |
85% | |
MFCD00005043 | |
Acid Red 94, C.I. 45440 | |
DPBQSRZQYAYDGY-UHFFFAOYSA-N | |
C1=C2C(=C(C(=C1I)O)I)OC3=C(C(=C(C=C3C24C5=C(C(=C(C(=C5Cl)Cl)Cl)Cl)C(=O)O4)I)O)I.[Na].[Na] | |
1019.644 | |
544 to 550nm | |
85% | |
Brown to Red | |
Powder |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Warning
- Rose Bengal
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H319-Causes serious eye irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only