missing translation for 'onlineSavingsMsg'
Learn More

ROCK1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 20247-1-AP-150UL

 View more versions of this product

Product Code. 16889764

  • 359.47 € / 150µL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

The antibody is specific to ROCK1.

The Rho target protein, Rho-associated coiled coil-containing protein kinase (ROCK)-1 has been found to be a new Caspase-3 substrate. ROCK, one of the effectors of the small GTPase Rho, has recently been shown to contribute significantly to myosin light chain (MLC) activation through two pathways: direct phosphorylation of MLC and phosphorylation of MLC phosphatase, leading to its inhibition. The increase in cellular contractility that is necessary for apoptotic membrane blebbing implies sustained augmentation of MLC phosphorylation. ROCK-1 consists of an amino-terminal kinase domain and an inhibitory cysteine/histidine-rich C-terminal domain that is located within a pleckstrin-homology region. They are joined by a variable region that contains the Rho-binding domain. During apoptosis, ROCK-I is cleaved by caspase-3 at a conserved DETD1113/G sequence and its carboxy-terminal inhibitory domain is removed, resulting in deregulated and constitutive kinase activity. The caspase-3-mediated cleavage and activation of ROCK I induces phosphorylation of MLC and membrane blebbing, one of the first events in the execution phase of apoptosis.
Specifications

Specifications

ROCK1
Polyclonal
Unconjugated
ROCK1
p160 ROCK 1, P160ROCK, PRO0435, ROCK1
Rabbit
Antigen Affinity Chromatography
RUO
19877, 6093, 81762
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.39 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P70335, Q13464, Q63644
ROCK1
Peptide
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers