missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ROBO4 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20221-1-AP-20UL
This item is not returnable.
View return policy
Description
This antibody is specific to ROBO4.
Roundabouts (ROBO) are cell-surface receptors that mediate repulsive signaling mechanisms at the central nervous system midline. However, ROBOs may also mediate attraction mechanisms in the context of vascular development. ROBO4 is a novel roundabout protein which is restricted in expression to endothelial cells in vitro and sites of angiogenesis in vivo. Because it is not expressed on normal endothelial cells, in vivo, it is a promising tumor endothelial marker.Specifications
ROBO4 | |
Polyclonal | |
Unconjugated | |
ROBO4 | |
ECSM4, Magic roundabout, MRB, ROBO4, Roundabout homolog 4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
54538, 74144 | |
-20°C | |
Liquid |
Western Blot | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8C310, Q8WZ75 | |
ROBO4 | |
Peptide | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |