Learn More
Macherey-Nagel™ RNaseA
Intended primarily for use with NucleoSpin™ and NucleoBond™ kits. Macherey-Nagel™ RNase A is a freeze-dried enzyme sourced from bovine pancreas. It has RNase activity of 120 U/mg. Available in 50mg and 100mg packs.
Brand: Macherey-Nagel™ 740505
Description
Ribonuclease (RNase) is an enzyme that catalyzes the degradation of RNA into smaller components. RNase A is one of the hardiest enzymes in common laboratory usage. It is specific for single-stranded RNAs, cleaving the 3'-end of unpaired C and U residues, and ultimately forming a 3'-phosphorylated product via a 2',3'-cyclic monophosphate intermediate. It does not require any cofactors for its activity.
- CAS: 9001-99-4
- For research purposes only
- For use, dissolve in water or resuspension buffer
- Dissolved in water, it can be stored at 4°C for up to 3 months (in smaller aliquots, at −20°C, for longer)
- Dissolved in resuspension buffer A1 / S1 / S1-EF / RES / RESEF / RES-BAC, it can be stored at 4°C for up to 6 months
Specifications
White | |
A1 / S1 / S1-EF / RES / RESEF / RES-BAC |
|
Bovine pancreas | |
Ribonuclease A |
NucleoSpin™ and NucleoBond™ kits | |
Store at room temperature (18–25°C) for at least 1 year | |
100 mg | |
Freeze-dried (Lyophilized) |
Safety and Handling
- Danger
Hazard Category
- Respiratory sensitiser Category 1
Hazard Statement
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Your input is important to us. Please complete this form to provide feedback related to the content on this product.