missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RIMS4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20091-1-AP-150UL
This item is not returnable.
View return policy
Description
The antibody is specific to RIMS4.
Regulates synaptic membrane exocytosis.Specifications
RIMS4 | |
Polyclonal | |
Unconjugated | |
RIMS4 | |
C20orf190, Rab3 interacting molecule 4, RIM 4, RIM4, RIM4 gamma, RIMS4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
140730, 241770, 266976 | |
-20°C | |
Liquid |
Western Blot | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P60191, Q8CIX1, Q9H426 | |
RIMS4 | |
Peptide | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction