Learn More
Rifampicin, Fisher BioReagents™
35.85 € - 437.00 €
Chemical Identifiers
CAS | 13292-46-1 |
---|---|
Molecular Formula | C43H58N4O12 |
Molecular Weight (g/mol) | 822.953 |
InChI Key | FZYOVNIOYYPUPY-HRJPTAQKSA-N |
Synonym | Rif |
PubChem CID | 131839595 |
SMILES | CC1C=CC=C(C(=O)NC2=C(C3=C(C(=C4C(=C3C(=O)C2=CNN5CCN(CC5)C)C(=O)C(O4)(OC=CC(C(C(C(C(C(C1O)C)O)C)OC(=O)C)C)OC)C)C)O)O)C |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10103443
|
Fisher BioReagents
BP2679-250 |
250 mg | Amber Glass | ||||||
10533325
|
Fisher BioReagents
BP2679-1 |
1 g | Amber Glass | ||||||
10396743
|
Fisher BioReagents
BP2679-5 |
5 g | Amber Glass | ||||||
10562975
|
Fisher BioReagents
BP267925 |
25 g | Amber Glass | ||||||
Description
Rifampicin
Rifampicin is a rifamycin which specifically inhibits DNA-dependent bacterial RNA Polymerase. Mammalian RNA polymerase is not affected.Chemical Identifiers
13292-46-1 | |
822.953 | |
Rif | |
CC1C=CC=C(C(=O)NC2=C(C3=C(C(=C4C(=C3C(=O)C2=CNN5CCN(CC5)C)C(=O)C(O4)(OC=CC(C(C(C(C(C(C1O)C)O)C)OC(=O)C)C)OC)C)C)O)O)C |
C43H58N4O12 | |
FZYOVNIOYYPUPY-HRJPTAQKSA-N | |
131839595 |
Specifications
Rifampicin | |
13292-46-1 | |
97% | |
C43H58N4O12 | |
Rif | |
CC1C=CC=C(C(=O)NC2=C(C3=C(C(=C4C(=C3C(=O)C2=CNN5CCN(CC5)C)C(=O)C(O4)(OC=CC(C(C(C(C(C(C1O)C)O)C)OC(=O)C)C)OC)C)C)O)O)C | |
131839595 | |
Orange |
Solid | |
183°C | |
< 1.0% | |
15, 8341 | |
FZYOVNIOYYPUPY-HRJPTAQKSA-N | |
822.953 | |
822.96 | |
Rifampicin |
Safety and Handling
WARNING!
Emergency Overview
Substances known to cause developmental toxicity in humans. Harmful if swallowed. May cause skin, eye, and respiratory tract irritation. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required.
NFPA
Health:2
Flammability:0
Instability:0
Recommended Storage : 4°C
- Rifampin
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
Precautionary Statement
- P270-Do not eat, drink or smoke when using this product.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.