Learn More
Thermo Scientific™ Richard-Allan Scientific™ Unbuffered Zinc Formalin
Achieve similar results to 10% formalin with Thermo Scientific™ Richard-Allan Scientific™ Unbuffered Zinc Formalin.
Brand:  Thermo Scientific™ 7600
Product Code. 10600984
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
Achieve similar results to 10% formalin with Thermo Scientific™ Richard-Allan Scientific™ Unbuffered Zinc Formalin.
  • Formalin does not contain mercury.
  • Easy-to-pour container minimizes splashing and reduces spills.