missing translation for 'onlineSavingsMsg'
Learn More

Richard-Allan Scientific™ Formaldehyde (37%), Epredia™

Fix and store tissue specimens in histological applications with Epredia™ Richard-Allan Scientific™ Formaldehyde (37%).

Brand:  Epredia 9311


Product Code. 12590007

This item is not returnable. View return policy

Description

Description

Easy-to-pour container minimizes splashing and reduces spills.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

1 gal.
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger

Hazard Category
  • Carcinogenicity Category 1A
  • Flammable liquid Category 3
  • Acute toxicity Category 3
  • Specific target organ toxicity Category 1
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Skin sensitiser Category 1
  • Specific target organ toxicity after single exposure Category 1

Hazard Statement
  • H226-Flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H350-May cause cancer.
  • H370-Causes damage to organs.

Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P271-Use only outdoors or in a well-ventilated area.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P235-Keep cool.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P363-Wash contaminated clothing before reuse.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P330-Rinse mouth.
  • P331-Do NOT induce vomiting.
  • P370+P378-In case of fire: Use CO2, dry chemical, or foam to extinguish.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : formaldehyde, methylalcohol

SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Richard-Allan Scientific™ Formaldehyde (37%), Epredia™ > 1 gal.

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.