missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ribavirin, 98%, Thermo Scientific Chemicals
Ribavirin, CAS# 36791-04-5, is an antiviral agent and a guanosine analog, which stops viral RNA synthesis and mRNA capping.
82.80 € - 571.00 €
Chemical Identifiers
CAS | 36791-04-5 |
---|---|
Molecular Formula | C8H12N4O5 |
Molecular Weight (g/mol) | 244.2 |
InChI Key | IWUCXVSUMQZMFG-AFCXAGJDSA-N |
Synonym | ribavirin, tribavirin, virazole, rebetol, ribavirine, copegus, vilona, ribamide, ribamidil, ribasphere |
PubChem CID | 37542 |
ChEBI | CHEBI:63580 |
IUPAC Name | 1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2,4-triazole-3-carboxamide |
SMILES | C1=NC(=NN1C2C(C(C(O2)CO)O)O)C(=O)N |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15599513
|
Thermo Scientific Acros
460480010 |
1 g |
82.80 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15509523
|
Thermo Scientific Acros
460480050 |
5 g |
227.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15519523
|
Thermo Scientific Acros
460480250 |
25 g |
571.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General description
• Ribavirin is a broad-spectrum antiviral nucleoside analog
• It inhibits viral genome synthesis
Applications
• Ribavirin is an antiviral compound
• Its metabolite, ribavirin 5'-phosphate, inhibits inosine monophosphate dehydrogenase (IMP), which is involved in GTP biosynthesis
Chemical Identifiers
36791-04-5 | |
244.2 | |
ribavirin, tribavirin, virazole, rebetol, ribavirine, copegus, vilona, ribamide, ribamidil, ribasphere | |
CHEBI:63580 | |
C1=NC(=NN1C2C(C(C(O2)CO)O)O)C(=O)N |
C8H12N4O5 | |
IWUCXVSUMQZMFG-AFCXAGJDSA-N | |
37542 | |
1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2,4-triazole-3-carboxamide |
Specifications
36791-04-5 | |
0.5% max. | |
97.5% min. (HPLC) | |
C8H12N4O5 | |
Solubility in water: 142 mg/mL @25°C. | |
C1=NC(=NN1C2C(C(C(O2)CO)O)O)C(=O)N | |
244.2 | |
CHEBI:63580 | |
98% |
0.25% max. | |
Authentic | |
Glass Bottle | |
ribavirin, tribavirin, virazole, rebetol, ribavirine, copegus, vilona, ribamide, ribamidil, ribasphere | |
IWUCXVSUMQZMFG-AFCXAGJDSA-N | |
1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2,4-triazole-3-carboxamide | |
37542 | |
244.2 | |
Ribavirin |
Safety and Handling
May damage fertility or the unborn child
Reproductive toxicity (category 1B)
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Ribavirin
Signal Word
- Danger
Hazard Category
- Reproductive toxicity Category 1B
Hazard Statement
- H360-May damage fertility or the unborn child.
Precautionary Statement
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
RUO – Research Use Only