missing translation for 'onlineSavingsMsg'
Learn More

Rhodium(II) acetate dimer, dihydrate, ca 43% Rh, Thermo Scientific Chemicals

298.49 € - 1157.76 €

Chemical Identifiers

CAS 29998-99-0
Molecular Formula C8H16O10Rh2
Molecular Weight (g/mol) 478.02
MDL Number MFCD00149212
Synonym Tetrakis[micron-(acetato-O:O')]diaquadirhodium
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10062580
SDS View Documents
Thermo Scientific Acros
197612500
250 mg Glass bottle
298.49 €
250mg
Please to purchase this item. Need a web account? Register with us today!
10402781
SDS View Documents
Thermo Scientific Acros
197610010
1 g Glass bottle
1157.76 €
1g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Rhodium(II) acetate dimer, dihydrate, ca 43% Rh, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.