missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rhodium, 5% on carbon paste, Type 594, Thermo Scientific™
Chemical Identifiers
CAS | 7440-16-6 |
---|---|
Molecular Formula | Rh |
Molecular Weight (g/mol) | 102.91 |
MDL Number | MFCD00011201 MFCD03458392 |
InChI Key | MHOVAHRLVXNVSD-UHFFFAOYSA-N |
Synonym | fume, on carbon, rhodium, elemental, rodio, black, rhodium, metal, unii-dmk383dsac, on alumina, rh/al2o3, dmk383dsac |
PubChem CID | 23948 |
ChEBI | CHEBI:33359 |
IUPAC Name | rhodium |
SMILES | [Rh] |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
7440-16-6 | |
102.91 | |
MHOVAHRLVXNVSD-UHFFFAOYSA-N | |
23948 | |
rhodium |
Rh | |
MFCD00011201 MFCD03458392 | |
fume, on carbon, rhodium, elemental, rodio, black, rhodium, metal, unii-dmk383dsac, on alumina, rh/al2o3, dmk383dsac | |
CHEBI:33359 | |
[Rh] |
Specifications
7440-16-6 | |
Solid | |
MFCD00011201 MFCD03458392 | |
MHOVAHRLVXNVSD-UHFFFAOYSA-N | |
rhodium | |
23948 | |
Odorless |
Gray to Black | |
Rh | |
fume, on carbon, rhodium, elemental, rodio, black, rhodium, metal, unii-dmk383dsac, on alumina, rh/al2o3, dmk383dsac | |
[Rh] | |
102.91 | |
CHEBI:33359 | |
Rhodium, 5% on carbon paste, Type 594 |
Safety and Handling
P210-P240-P241-P280-P370+P378q
H228
missing translation for 'einecsNumber' : 231-125-0
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures