Learn More
Rhenium powder, -22 mesh, Puratronic™, 99.997% (metals basis), Thermo Scientific Chemicals
102.00 € - 326.00 €
Chemical Identifiers
CAS | 7440-15-5 |
---|---|
Molecular Formula | Re |
Molecular Weight (g/mol) | 186.21 |
MDL Number | MFCD00011195 |
InChI Key | WUAPFZMCVAUBPE-UHFFFAOYSA-N |
Synonym | rhenium, elementar, renio, unii-7yhu292iny, 7yhu292iny, foil, ion re7, rhenium, ion re7, atom, hydride, pellets |
PubChem CID | 23947 |
ChEBI | CHEBI:49882 |
IUPAC Name | rhenium |
SMILES | [Re] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11310909
|
Thermo Scientific Alfa Aesar
010673.03 |
1 g |
102.00 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11320909
|
Thermo Scientific Alfa Aesar
010673.06 |
5 g |
326.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7440-15-5 | |
186.21 | |
WUAPFZMCVAUBPE-UHFFFAOYSA-N | |
23947 | |
rhenium |
Re | |
MFCD00011195 | |
rhenium, elementar, renio, unii-7yhu292iny, 7yhu292iny, foil, ion re7, rhenium, ion re7, atom, hydride, pellets | |
CHEBI:49882 | |
[Re] |
Specifications
7440-15-5 | |
MFCD00011195 | |
rhenium, elementar, renio, unii-7yhu292iny, 7yhu292iny, foil, ion re7, rhenium, ion re7, atom, hydride, pellets | |
[Re] | |
186.21 | |
CHEBI:49882 | |
Puratronic™ | |
Powder | |
Rhenium powder |
Re | |
UN3089 | |
WUAPFZMCVAUBPE-UHFFFAOYSA-N | |
rhenium | |
23947 | |
99.999% | |
−22 Mesh | |
(metals basis) |
Safety and Handling
GHS H Statement
H228
Flammable solid.
GHS P Statement
P210-P280-P240-P241-P370+P378a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
Use explosion-proof electrical/ventilating/lighting/ equipment.
In case of fire: Use for extinction: CO2, powder or water spray.
Warning
missing translation for 'dotInformation' : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL POWDERS, FLAMMABLE, N.O.S.
missing translation for 'einecsNumber' : 231-124-5
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only