Learn More
Rhenium(IV) sulfide, 99%, Thermo Scientific Chemicals
344.72 € - 1596.95 €
Chemical Identifiers
CAS | 12038-63-0 |
---|---|
Molecular Formula | ReS2 |
Molecular Weight (g/mol) | 250.33 |
MDL Number | MFCD00016307 |
InChI Key | USWJSZNKYVUTIE-UHFFFAOYSA-N |
Synonym | rhenium sulfide, rhenium disulphide, disulfanylidenerhenium, dithioxorhenium, rhenium iv sulfide, bis sulfanylidene rhenium |
PubChem CID | 82864 |
IUPAC Name | bis(sulfanylidene)rhenium |
SMILES | S=[Re]=S |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11433790
|
Thermo Scientific Alfa Aesar
089482.04 |
2 g |
344.72 €
2g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11423790
|
Thermo Scientific Alfa Aesar
089482.09 |
10 g |
1596.95 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Rhenium(IV) sulfide is used as a diamagnetic semiconductor.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
12038-63-0 | |
250.33 | |
USWJSZNKYVUTIE-UHFFFAOYSA-N | |
82864 | |
S=[Re]=S |
ReS2 | |
MFCD00016307 | |
rhenium sulfide, rhenium disulphide, disulfanylidenerhenium, dithioxorhenium, rhenium iv sulfide, bis sulfanylidene rhenium | |
bis(sulfanylidene)rhenium |
Specifications
12038-63-0 | |
ReS2 | |
rhenium sulfide, rhenium disulphide, disulfanylidenerhenium, dithioxorhenium, rhenium iv sulfide, bis sulfanylidene rhenium | |
USWJSZNKYVUTIE-UHFFFAOYSA-N | |
bis(sulfanylidene)rhenium | |
82864 | |
99% | |
7.506 g/mL |
Powder | |
MFCD00016307 | |
Insoluble in water. Soluble in 1N sodium hydroxide solution. | |
S=[Re]=S | |
250.33 | |
250.33 | |
Odorless | |
Rhenium (IV) sulfide |
Safety and Handling
missing translation for 'einecsNumber' : 234-878-3
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.