Learn More
Rhenium(III) chloride, Thermo Scientific Chemicals
156.00 € - 416.00 €
Chemical Identifiers
CAS | 13569-63-6 |
---|---|
Molecular Formula | Cl3Re |
Molecular Weight (g/mol) | 292.557 |
MDL Number | MFCD00011196 |
InChI Key | LOIHSHVELSAXQN-UHFFFAOYSA-K |
Synonym | rhenium iii chloride, rhenium trichloride, rhenium chloride recl3, rhenium 3+ trichloride, recl3, acmc-20ajss |
PubChem CID | 83581 |
IUPAC Name | trichlororhenium |
SMILES | Cl[Re](Cl)Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11366678
|
Thermo Scientific Alfa Aesar
013230.01 |
0.25 g |
156.00 €
0.25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11356678
|
Thermo Scientific Alfa Aesar
013230.03 |
1 g |
416.00 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It is used as a starting material for synthesis of rhenium complexes. ReCl3 can react with peralkylated benzene-metal complexes that act as electron reservoirs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
13569-63-6 | |
292.557 | |
LOIHSHVELSAXQN-UHFFFAOYSA-K | |
83581 | |
Cl[Re](Cl)Cl |
Cl3Re | |
MFCD00011196 | |
rhenium iii chloride, rhenium trichloride, rhenium chloride recl3, rhenium 3+ trichloride, recl3, acmc-20ajss | |
trichlororhenium |
Specifications
13569-63-6 | |
Powder | |
MFCD00011196 | |
Soluble in water. | |
Cl[Re](Cl)Cl | |
292.557 | |
292.56 | |
Moisture sensitive | |
Rhenium(III) chloride |
Red | |
Cl3Re | |
rhenium iii chloride, rhenium trichloride, rhenium chloride recl3, rhenium 3+ trichloride, recl3, acmc-20ajss | |
LOIHSHVELSAXQN-UHFFFAOYSA-K | |
trichlororhenium | |
83581 | |
Odorless | |
Dark red powder |
Safety and Handling
In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable gloves.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 236-987-1
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only