missing translation for 'onlineSavingsMsg'
Learn More

Rhenium foil, 0.5mm (0.02in) thick, annealed, 99.97% (metals basis), Thermo Scientific Chemicals

568.83 € - 1679.36 €

Chemical Identifiers

CAS 7440-15-5
Molecular Formula Re
Molecular Weight (g/mol) 186.21
MDL Number MFCD00011195
InChI Key WUAPFZMCVAUBPE-UHFFFAOYSA-N
Synonym rhenium, elementar, renio, unii-7yhu292iny, 7yhu292iny, foil, ion re7, rhenium, ion re7, atom, hydride, pellets
PubChem CID 23947
ChEBI CHEBI:49882
IUPAC Name rhenium
SMILES [Re]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11331499
SDS View Documents
Thermo Scientific Alfa Aesar
013983.FF
25 x 25 mm
568.83 €
25mm
Please to purchase this item. Need a web account? Register with us today!
11450390
SDS View Documents
Thermo Scientific Alfa Aesar
013983.FI
50 x 50 mm
1679.36 €
Each
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7440-15-5
186.21
WUAPFZMCVAUBPE-UHFFFAOYSA-N
23947
rhenium
Re
MFCD00011195
rhenium, elementar, renio, unii-7yhu292iny, 7yhu292iny, foil, ion re7, rhenium, ion re7, atom, hydride, pellets
CHEBI:49882
[Re]

Specifications

7440-15-5
MFCD00011195
WUAPFZMCVAUBPE-UHFFFAOYSA-N
rhenium
23947
99.97%
≈6.58g/25x25mm
(metals basis)
Re
rhenium, elementar, renio, unii-7yhu292iny, 7yhu292iny, foil, ion re7, rhenium, ion re7, atom, hydride, pellets
[Re]
186.21
CHEBI:49882
Odorless
Foil
Rhenium foil, 0.5mm (0.02 in.) thick, Annealed
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 231-124-5

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Rhenium foil, 0.5mm (0.02in) thick, annealed, 99.97% (metals basis), Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.