Learn More
Thermo Scientific™ Revco™ Plasma Freezers
Simplify operation and increase accuracy with these plasma freezers, designed to meet AABB, ANRC and FDA standards for safety and performance.
Description
Simplify operation and increase accuracy with the microprocessor control of Thermo Scientific™ Revco™ Plasma Freezers. Our −30°C Plasma Freezers are designed to meet AABB, ANRC and FDA standards for safety and performance in plasma storage. Five models offer cabinet sizes to meet a variety of space needs and storage volume requirements.
Key Features- CFC-free refrigerant
- Preset temperature at -30°C
- Automatic condensate removal and defrost, with sensor that automatically initiates defrost cycle on demand, eliminating need for a defrost timer, minimizing frost build-up and optimizing coil efficiency
- Positive airflow systems
Microprocessor Control
- Icon-based controls and displays
- Preset temperature setpoint
- Graphic thermometer is steady green when temperature is normal, flashes top bar for overtemperature and flashes bulb for undertemperature
- Large, easy-to-read digital display shows temperature within 1°C
- Audible/visual warnings for overtemperature, undertemperature or power failure
- Key-operated OFF/ON/ALARM ON master switch for setpoint security
- Push-button alarm test
- Remote alarm contacts
- Alarm silence, ringback and auto reset
- Controller battery backup system
Construction
- Cold-rolled steel cabinet surfaces finished with high-impact powder paint coating
- One solid door, hinged on right (except 51.1 cu. ft. model: two solid doors with keyed lock, self-close against closed-cell peripheral gasket)
- Full-length door handles
- Adjustable leveling feet
- High-density, CFC-free urethane foam insulation protects cabinet interior from ambient temperature fluctuations, minimizes operating costs
- Positive, forced-air circulation maintains uniformity at all shelf levels
- Directional airflow assures quick recovery after door openings
- Heavy-duty, hermetically sealed compressors
- 1 in. (2.5cm) access port with plug on undercounter model only
Ordering Information:
Alarm systems, temperature chart recorders and power monitors available separately. For additional voltages, contact your CSR.
Includes:
Space-saver and upright models come with a 6 in. (15.2cm) seven-day circular chart temperature recorder (optional free-standing on undercounter model) and extra-long probe for manual alarm testing.
Warranty varies by country, please contact us for detailsYour input is important to us. Please complete this form to provide feedback related to the content on this product.