Learn More
Trajan Scientific and Medical™ Replacement Needles for Luer-Lock Syringes
Range of quality needles to fit a SGE syringe in a range of lengths and tip styles.
Brand: Trajan Scientific and Medical™ 039847
This item is not returnable.
View return policy
Description
- Needle length is the exposed length of needle
- Choose a different replacement needle by selecting syringe volume, needle length, OD and tip style
Specifications
Borosilicate Glass (Barrel) | |
0.80 mm | |
Replacement Needles for Luer-Lock Syringes | |
7 cm |
18 G | |
1.27 mm | |
5/Pk. | |
Bevel |