Learn More
Trajan Scientific and Medical™ Replacement Needles for Agilent Syringes
Range of quality needles to fit a SGE syringe in a range of lengths and tip styles.
Brand:  Trajan Scientific and Medical™ 038717
Product Code. 15218603
  • 40.70 € / Pack of 2
Explore more special offers
Description
Description
  • Needle length is the exposed length of needle
  • Choose a different replacement needle by selecting syringe volume, needle length, OD and tip style
Specifications
Specifications
23 G
0.63 mm
Cone
0.24 mm
50 μL
Agilent Syringes