Learn More
Thermo Scientific™ Remel™ Agglutinating Sera, E. coli Type O
Used for serological identification of E. coli by slide and/or tube agglutination tests.
Specifications
Quantity 2 mL
View More Specs
Description
Description
Remel Agglutinating Sera, E. coli Type O is used for serological identification of E. coli by slide and/or tube agglutination tests.
Specifications
Specifications
2 mL