missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Refractive Index Certified Reference Material at 20 °C, 25 °C and 30 °C (Nominal RI Value 1.6556 at 25 °C), UKAS ISO 17025 / ISO 17034 Certified, in Tamper-Evident Glass Vials, 5 x 10 mL, Paragon Scientific™
Paragon Scientific’s range of Refractive Index Certified Reference Materials (CRMs) provide certified values for refractive index measurements at 20 °C, 25 °C and 30 °C and cover a wide range of calibration needs.
Brand: Paragon Scientific PS-RI-10K
This item is not returnable.
View return policy
Description
- Refractive Index Certified Reference Materials at 20 °C, 25 °C and 30 °C
- Certified in strict accordance to ISO 17025 and ISO 17034 under our UKAS accreditation
- Highest level of accreditation guarantee, providing the most credible certified data available worldwide
- All measurements are fully traceable to NIST and international protocols
- Low levels of uncertainty, ensuring maximum accuracy of data at hand and dependable results
- Available in single 10 mL volume tamper-evident glass bottles or as a multi-pack of 5 per material (includes set of disposable pipettes)
- 12 month shelf life
- Manufactured in the United Kingdom
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Refractive Index Certified Reference Material at 20°C, 25°C and 30°C (Nominal RI Value1.6556 at 25°C)
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H302-Harmful if swallowed.
- H319-Causes serious eye irritation.
Precautionary Statement
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : 1-Bromonaphthalene
Nominal values quoted. Certified Values will vary from batch to batch.