Learn More
3M™ Refine 300 Series Anti-Scratch, Anti-Fog Safety Glasses
Provides excellent coverage with fewer gaps where dangerous flying debris can damage the eyes. 3M™ Refine 300 Series Anti-Scratch, Anti-Fog Safety Glasses are especially designed to fit smaller head and face shapes.
12.88 € - 16.43 €
Specifications
Lens Coating | Anti-fog, Scratch-resistant |
---|---|
Product Type | Safety glasses; Safety Spectacles |
No. per Pack | 1 Ea. |
Lens Material | Polycarbonate |
Description
- Available in 2 lens colors
- Slim frame design for younger or female users
- Integrated side shield for added protection
- Soft, padded adjustable nose bridge provides a comfortable fit
Specifications
Anti-fog, Scratch-resistant | |
Safety glasses; Safety Spectacles | |
1 Ea. | |
Polycarbonate |