Learn More
Lenz Laborglasintrumente™ Reduction Adapters with Ground Joints
Adapter for conical ground sockets and cones made of DURAN™ tubing
15.00 € - 75.00 €
Specifications
Material | Duran Tubing |
---|---|
Product Type | Reduction adapter with conical ground sockets and cones |
Quantity | 1 |
Product Code | Brand | Cone Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Cone Size | Price | Quantity & Availability | |||||
10131901
|
Lenz Laborglasintrumente™
01501424 |
24/29 |
15.00 €
Each |
Estimated Shipment: 22-01-2025
Log in to see stock. |
|||||
10264571
|
Lenz Laborglasintrumente™
01502434 |
34/35 |
33.00 €
Each |
Estimated Shipment: 07-02-2025
Log in to see stock. |
|||||
10070341
|
Lenz Laborglasintrumente™
01502960 |
60/46 |
75.00 €
Each |
Estimated Shipment: 22-01-2025
Log in to see stock. |
|||||
Description
- Conical ground sockets and cones
- Made of DURAN™ tubing
Specifications
Duran Tubing | |
Reduction adapter with conical ground sockets and cones | |
1 |