missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human TFB2M Protein

A cDNA sequence encoding the TFB2M was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13728-10ug

 View more versions of this product

Product Code. 15984509

  • 222.00 € / 10µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

TFB2M Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMAGRFCI LGSEAATRKH LPARNHCGLS DSSPQLWPEP DFRNPPRKAS KASLDFKRYV TDRRLAETLA QIYLGKPSRP PHLLLECNPG PGILTQALLE AGAKVVALES DKTFIPHLES LGKNLDGKLR VIHCDFFKLD PRSGGVIKPP AMSSRGLFKN LGIEAVPWTA DIPLKVVGMF PSRGEKRALW KLAYDLYSCT SIYKFGRIEV NMFIGEKEFQ KLMADPGNPD LYHVLSVIWQ LACEIKVLHM EPWSSFDIYT RKGPLENPKR RELLDQLQQK LYLIQMIPRQ NLFTKNLTPM NYNIFFHLLK HCFGRRSATV IDHLRSLTPL DARDILMQIG KQEDEKVVNM HPQDFKTLFE TIERSKDCAY KWLYDETLED R
Greater than 90.0% as determined by SDS-PAGE.
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
TFB2M protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 10% glycerol and 1mM DTT.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers