missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MEOX2 Protein
A cDNA sequence encoding the MEOX2 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12677-2ug
This item is not returnable.
View return policy
Specifications
MEOX2 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSRKERTAF TKEQIRELEA EFAHHNYLTR LRRYEIAVNL DLTERQVKVW FQNRRMKWKR VKGGQQGAAA REKELVNVKK GTLLPSELSG IGAATLQQTG DSIANEDSHD SDHSSEHAHL | |
Greater than 95% as determined by SDS-PAGE. |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The MEOX2 solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.2M NaCl, 40% glycerol and 2mM DTT. |