Learn More
Macherey-Nagel™ Receiver Plates
Macherey-Nagel 96-well receiver plates with inserted filter frits of 35 μm pore size are suitable for centrifugation and vacuum applications for high throughput work.
Brand:  Macherey-Nagel™ 740512.4
200.95 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 16246551
  • 238.00 € / Pack of 4
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • 96-well plates
  • For centrifugation and vacuum
  • 35 μm filter frits
Specifications
Specifications
96
35 μL
Suitable for Centrifugation and Vacuum Use
Filter Frits