Learn More
Macherey-Nagel™ Receiver Plates
Macherey-Nagel 96-well receiver plates with inserted filter frits of 35 μm pore size are suitable for centrifugation and vacuum applications for high throughput work.
Brand:  Macherey-Nagel™ 740513.4
 View more versions of this product
Product Code. 16183547
  • 198.00 € / Pack of 4
Estimated Shipment: 20-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • 96-well plates
  • For centrifugation and vacuum
  • 35 μm filter frits
Specifications
Specifications
96
35 μL
Suitable for Gravity Flow, Centrifugation, and Vacuum Use
Hydrophilised Filter Frits