Learn More
RBP3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 14352-1-AP-20UL
 View more versions of this product
Product Code. 16852954
  • 145.00 € / 20µL
Explore more special offers
Description
Description
IRBP shuttles 11-cis and all trans retinoids between the retinol isomerase in the pigment epithelium and the visual pigments in the photoreceptor cells of the retina.
Specifications
Specifications
RBP3
Polyclonal
Unconjugated
RBP3
D10S64, D10S65, D10S66, IRBP, RBP3, RBPI, Retinol binding protein 3
Rabbit
Antigen Affinity Chromatography
RUO
19661, 24711, 5949
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation
0.4 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P10745, P49194
RBP3
RBP3 Fusion Protein Ag5699
20 μL
Primary
Mouse, Rat, Human