missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IFN gamma Rat ELISA Kit

Measure levels of rat interferon gamma (IFNg) in rat serum, plasma (EDTA, heparin, sodium citrate), and culture supernatant samples using 96-well plates.

672.00 € - 980.00 €

Specifications

Accession Number P01581
Assay Range 8 to 500pg/mL
Assay Sensitivity <2pg/mL
Description Rat IFN gamma ELISA Kit
Conjugate Biotin
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10077003
View Documents
Invitrogen™
ERIFNG
96 Tests
672.00 €
1 set
Please to purchase this item. Need a web account? Register with us today!
10750625
View Documents
Invitrogen™
ERIFNG2
2 x 96 Tests
980.00 €
1 set
Estimated Shipment: 09-01-2025
to see stock.
Add to basket
 
Description

Description

Assay IFN-γ(rat interferon gamma) levels in rat serum, plasma (EDTA, heparin, sodium citrate), and culture supernatant samples with our Thermo Scientific Rat IFN gamma ELISA Kits.

These rat IFN-gamma ELISA kits use pre-titered, matched pairs of coating and detection antibodies to achieve sensitive and accurate measurement of interferon gamma (IFNγ) in samples at 50 per well. Also included are calibrated recombinant protein standard, streptavidin-HRP, TMB substrate solution and stop solution. The complete kits contain precoated 96-well strip plate(s) and lot-optimized reagents. All components undergo quality control testing to minimize lot-to-lot variability.
  • Target: Natural and recombinant rat interferon gamma (IFNγ)

  • Format: Colorimetric ELISA (TMB substrate) in clear 96-well strip plate

  • Measurement: Absorbance at 450nm minus absorbance at 550nm

  • Assay Range: 8 to 500pg/mL

  • Sensitivity: < 2pg/mL

  • Sample Types: Cell culture supernatant, serum, plasma (EDTA, sodium citrate and heparin)

  • Total Assay Time: 2.5 hours

  • Sample Size: 50μL

  • Specificity: No significant cross-reaction with rat IL-1α, IL-1β, IL-2, IL-4, IL-6, IL-10, MIP-1α, RANTES, or TNFα, or human or pig IFNγ. When preparations of NIH/NIBSC mouse IFNγ (lot Gg02-901-533) were evaluated in the Rat IFNγ ELISA Kit, 6250pg/mL mouse IFNγ was equivalent to 258pg/mL rat IFNγ (4.1% cross-reactivity)

Includes:Complete kits contain pre-coated antibody-plates, detection antibodies, buffers, diluents, standards, and substrates.

Specifications

Specifications

P01581
<2pg/mL
Biotin
ELISA Kit
Colorimetric Microplate Reader
IFNG2
8.5%
Absorbance at 450nm minus absorbance at 550nm
Plasma,50μL; Serum,50μL; Supernatant,50μL
Rat
2 hr. 30 min.
8 to 500pg/mL
Rat IFN gamma ELISA Kit
Colorimetric ELISA (TMB substrate) with Plates
Cell Culture Supernatant, Plasma (EDTA, Citrate, Heparin), Serum
25712
8.7%
HRP
RUO
2°C to 8°C
1 hr. 20 min.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.