missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Raptor Rabbit anti-Human, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20984-1-AP-20UL
This item is not returnable.
View return policy
Description
The antibody is specific to RPTOR.
Regulatory associated protein of mTOR (RAPTOR) is a binding partner of target of rapamycin (TOR). It binds to 4EBP1 and p70alpha. The binding of raptor to mTOR is essential for the mTOR-catalyzed phosphorylation of 4EBP1 in vitro. It enhances the mTOR kinase activity toward p70alpha. It is also an essential scaffold for the mTOR-catalyzed phosphorylation of 4EBP1 and mediates TOR action in vivo.Specifications
Raptor | |
Polyclonal | |
Unconjugated | |
RPTOR | |
KIAA1303, raptor, RPTOR | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
287871, 57521 | |
-20°C | |
Liquid |
Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8N122 | |
Rptor | |
Peptide | |
20 μL | |
Primary | |
Rat, Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction