Learn More
Raptor Rabbit anti-Human, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 20984-1-AP-150UL
 View more versions of this product
Product Code. 16800575
  • 410.00 € / 150µL
Explore more special offers
Description
Description
The antibody is specific to RPTOR.
Regulatory associated protein of mTOR (RAPTOR) is a binding partner of target of rapamycin (TOR). It binds to 4EBP1 and p70alpha. The binding of raptor to mTOR is essential for the mTOR-catalyzed phosphorylation of 4EBP1 in vitro. It enhances the mTOR kinase activity toward p70alpha. It is also an essential scaffold for the mTOR-catalyzed phosphorylation of 4EBP1 and mediates TOR action in vivo.
Specifications
Specifications
Raptor
Polyclonal
Unconjugated
RPTOR
KIAA1303, raptor, RPTOR
Rabbit
Antigen Affinity Chromatography
RUO
287871, 57521
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.2 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8N122
Rptor
Peptide
150 μL
Primary
Human, Rat